Thulium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Thulium trinitrate hydrate
14579-04
14579-09
14579-18
CAAA14579-04EA
153.42
CAD
CAAA14579-04
CAAA14579-09
CAAA14579-18
Thulium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Thulium(III) nitrate hydrate
Purity: 99.9%
Molecular Formula: Tm(NO3)3.xH2O (x≈5)
Formula Weight: 354.95(anhy)
Physical Form: Crystalline Aggregates
UN No.: 1477
MDL No.: MFCD00149875
Formula:
N₃O₉Tm·xH₂O MW: 354.95 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00149875 CAS Number: 36548-87-5 EINECS: 239-069-9 UN: 1477 ADR: 5.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...