Thallium, Plasma Standard Solution, Specpure®, Tl 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14437-AD
14437-AE
14437-AP
CAAA14437-ADEA
104.12
CAD
CAAA14437-AD
CAAA14437-AE
CAAA14437-AP
Thallium, Plasma Standard Solution, Specpure®, Tl 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Thallium Standards
Molecular Formula: Tl in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134063
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...