Praseodymium, plasma standard solution, Specpure®, Pr 10,000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14433-AD
14433-AE
14433-AP
CAAA14433-ADEA
144.89
CAD
CAAA14433-AD
CAAA14433-AE
CAAA14433-AP
Praseodymium, plasma standard solution, Specpure®, Pr 10,000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Praseodymium Standards
Molecular Formula: Pr6O11 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011174
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...