Neodymium, Plasma Standard Solution, Specpure®, Nd 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14431-AD
14431-AE
14431-AP
CAAA14431-ADEA
161.16
CAD
CAAA14431-AD
CAAA14431-AE
CAAA14431-AP
Neodymium, Plasma Standard Solution, Specpure®, Nd 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Neodymium Standards
Molecular Formula: Nd2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011130
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...