Magnesium, Plasma Standard Solution, Specpure®, Mg 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14430-AD
14430-AE
14430-AP
CAAA14430-ADEA
120.55
CAD
CAAA14430-AD
CAAA14430-AE
CAAA14430-AP
Magnesium, Plasma Standard Solution, Specpure®, Mg 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Magnesium Standards
Molecular Formula: Mg In 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085308
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...