Lanthanum, Plasma Standard Solution, Specpure®, La 10000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14429-AD
14429-AE
14429-AP
CAAA14429-ADEA
120.4
CAD
CAAA14429-AD
CAAA14429-AE
CAAA14429-AP
Lanthanum, Plasma Standard Solution, Specpure®, La 10000µg/ml, Thermo Scientific
Standards
Cationic Standards
Lanthanum Standards
Molecular Formula: La2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011066
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...