Gallium, plasma standard solution, Specpure®, Ga 10,000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14427-AD
14427-AE
14427-AP
CAAA14427-ADEA
137.93
CAD
CAAA14427-AD
CAAA14427-AE
CAAA14427-AP
Gallium, plasma standard solution, Specpure®, Ga 10,000µg/ml, Thermo Scientific
Standards
Cationic Standards
Gallium Standards
Molecular Formula: Ga in 5% HNO3/tr. HCl
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134045
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...