Dysprosium, plasma standard solution, Specpure®, Dy 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14426-AD
14426-AE
14426-AP
CAAA14426-ADEA
147.73
CAD
CAAA14426-AD
CAAA14426-AE
CAAA14426-AP
Dysprosium, plasma standard solution, Specpure®, Dy 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Dysprosium Standards
Molecular Formula: Dy2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010982
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...