Cesium, Plasma Standard Solution, Specpure®, Cs 10,000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14425-AD
14425-AE
14425-AP
CAAA14425-ADEA
153.42
CAD
CAAA14425-AD
CAAA14425-AE
CAAA14425-AP
Cesium, Plasma Standard Solution, Specpure®, Cs 10,000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Cesium Standards
Molecular Formula: Cs2CO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134037
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...