Yttrium, Plasma Standard Solution, Specpure®, Y 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14422-AD
14422-AE
14422-AP
CAAA14422-ADEA
127.51
CAD
CAAA14422-AD
CAAA14422-AE
CAAA14422-AP
Yttrium, Plasma Standard Solution, Specpure®, Y 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Yttrium Standards
Molecular Formula: Y2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011468
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...