Tantalum, Plasma Standard Solution, Specpure®, Ta 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14419-AD
14419-AE
14419-AP
CAAA14419-ADEA
140.3
CAD
CAAA14419-AD
CAAA14419-AE
CAAA14419-AP
Tantalum, Plasma Standard Solution, Specpure®, Ta 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Tantalum Standards
Molecular Formula: TaCl5 in 2% HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011252
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...