Silver, Plasma Standard Solution, Specpure®, Ag 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14418-AD
14418-AE
14418-AP
CAAA14418-ADEA
120.71
CAD
CAAA14418-AD
CAAA14418-AE
CAAA14418-AP
Silver, Plasma Standard Solution, Specpure®, Ag 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Silver Standards
Molecular Formula: Ag in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003397
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...