Samarium, plasma standard solution, Specpure®, Sm 10000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

14417-AD 14417-AE 14417-AP
CAAA14417-ADEA 137.93 CAD
CAAA14417-AD CAAA14417-AE CAAA14417-AP
Samarium, plasma standard solution, Specpure®, Sm 10000 µg/ml, Thermo Scientific
Standards Cationic Standards Samarium Standards

Molecular Formula: Sm2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011233

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR