Rhenium, Plasma Standard Solution, Specpure®, Re 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14416-AD
14416-AE
14416-AP
CAAA14416-ADEA
262.28
CAD
CAAA14416-AD
CAAA14416-AE
CAAA14416-AP
Rhenium, Plasma Standard Solution, Specpure®, Re 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Rhenium Standards
Molecular Formula: Re in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011195
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...