Nickel, Plasma Standard Solution, Specpure®, Ni 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14414-AD
14414-AE
14414-AP
CAAA14414-ADEA
121.34
CAD
CAAA14414-AD
CAAA14414-AE
CAAA14414-AP
Nickel, Plasma Standard Solution, Specpure®, Ni 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Nickel Standards
Molecular Formula: Ni in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011137
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...