Manganese, Plasma Standard Solution, Specpure®, Mn 10000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

14413-AD 14413-AE 14413-AP
CAAA14413-ADEA 126.4 CAD
CAAA14413-AD CAAA14413-AE CAAA14413-AP
Manganese, Plasma Standard Solution, Specpure®, Mn 10000 µg/ml, Thermo Scientific
Standards Cationic Standards Manganese Standards

Molecular Formula: Mn in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011111

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR