Germanium, plasma standard solution, Specpure®, Ge 10,000µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
14410-AD 14410-AE 14410-AE 14410-AP 14410-AD 14410-AP
CAAA14410-APEA 461.36 CAD
CAAA14410-AP AA14410-AP CAAA14410-AD CAAA14410-AE AA14410-AD AA14410-AE
Germanium, plasma standard solution, Specpure®, Ge 10,000µg/ml, Thermo Scientific
Standards Cationic Standards Germanium Standards
Molecular Formula: Ge in 5% HNO3/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085310
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR