Chromium, Plasma Standard Solution, Specpure®, Cr 10,000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14408-AD
14408-AE
14408-AP
CAAA14408-ADEA
116.76
CAD
CAAA14408-AD
CAAA14408-AE
CAAA14408-AP
Chromium, Plasma Standard Solution, Specpure®, Cr 10,000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Chromium Standards
Molecular Formula: Cr in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00010944
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...