Calcium, Plasma Standard Solution, Specpure®, Ca 10,000µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

14407-AD 14407-AE 14407-AP
CAAA14407-ADEA 122.45 CAD
CAAA14407-AD CAAA14407-AE CAAA14407-AP
Calcium, Plasma Standard Solution, Specpure®, Ca 10,000µg/ml, Thermo Scientific
Standards Cationic Standards Calcium Standards

Molecular Formula: CaCO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085314

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR