Zinc, Plasma Standard Solution, Specpure®, Zn 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14404-AD
14404-AE
14404-AP
CAAA14404-ADEA
127.51
CAD
CAAA14404-AD
CAAA14404-AE
CAAA14404-AP
Zinc, Plasma Standard Solution, Specpure®, Zn 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Zinc Standards
Molecular Formula: Zn in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011291
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...