Thulium, Plasma Standard Solution, Specpure®, Tm 10000 mg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14402-AD
14402-AE
CAAA14402-ADEA
172.22
CAD
CAAA14402-AD
CAAA14402-AE
CAAA14402-AP
Thulium, Plasma Standard Solution, Specpure®, Tm 10000 mg/ml, Thermo Scientific
Standards
Cationic Standards
Thulium Standards
Molecular Formula: Tm2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011281
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...