Platinum, Plasma Standard Solution, Specpure®, Pt 10,000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

14397-AD 14397-AE 14397-AP
CAAA14397-ADEA 595.66 CAD
CAAA14397-AD CAAA14397-AE CAAA14397-AP
Platinum, Plasma Standard Solution, Specpure®, Pt 10,000 µg/ml, Thermo Scientific

Molecular Formula: Pt in 20% HCl
Physical Form: Liquid
Odor: Characteristic
UN No.: 1789
MDL No.: MFCD00011179

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR