Lithium, Plasma Standard Solution, Specpure®, Li 10000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14394-AD
14394-AE
14394-AP
CAAA14394-ADEA
121.98
CAD
CAAA14394-AD
CAAA14394-AE
CAAA14394-AP
Lithium, Plasma Standard Solution, Specpure®, Li 10000µg/ml, Thermo Scientific
Standards
Cationic Standards
Lithium Standards
Molecular Formula: Li2CO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134051
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...