Gold, plasma standard solution, Specpure®, Au 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14392-AD
14392-AE
14392-AP
CAAA14392-ADEA
486.64
CAD
CAAA14392-AD
CAAA14392-AE
CAAA14392-AP
Gold, plasma standard solution, Specpure®, Au 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Gold Standards
Molecular Formula: Au in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00003436
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...