Cobalt, plasma standard solution, Specpure®, Co 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14390-AD
14390-AE
14390-AP
CAAA14390-ADEA
121.66
CAD
CAAA14390-AD
CAAA14390-AE
CAAA14390-AP
Cobalt, plasma standard solution, Specpure®, Co 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Cobalt Standards
Molecular Formula: Co in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010935
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...