Bismuth, Plasma Standard Solution, Specpure®, Bi 10,000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14388-AD
14388-AE
14388-AP
CAAA14388-ADEA
108.23
CAD
CAAA14388-AD
CAAA14388-AE
CAAA14388-AP
Bismuth, Plasma Standard Solution, Specpure®, Bi 10,000µg/ml, Thermo Scientific
Standards
Cationic Standards
Bismuth Standards
Molecular Formula: Bi in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134033
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...