Zirconium, Plasma Standard Solution, Specpure®, Zr 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14386-AD
14386-AE
14386-AP
CAAA14386-ADEA
153.89
CAD
CAAA14386-AD
CAAA14386-AE
CAAA14386-AP
Zirconium, Plasma Standard Solution, Specpure®, Zr 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Zirconium Standards
Molecular Formula: ZrCl2O in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011303
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...