Vanadium, Plasma Standard Solution, Specpure®, V 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14385-AD
14385-AE
14385-AP
CAAA14385-ADEA
124.19
CAD
CAAA14385-AD
CAAA14385-AE
CAAA14385-AP
Vanadium, Plasma Standard Solution, Specpure®, V 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Vanadium Standards
Molecular Formula: V2O5 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011453
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...