Tin, Plasma Standard Solution, Specpure®, Sn 10000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14384-AD
14384-AE
14384-AP
CAAA14384-ADEA
145.68
CAD
CAAA14384-AD
CAAA14384-AE
CAAA14384-AP
Tin, Plasma Standard Solution, Specpure®, Sn 10000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Tin Standards
Molecular Formula: Sn in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00133862
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...