Selenium, plasma standard solution, Specpure®, Se 10,000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

14381-AD 14381-AE 14381-AP
CAAA14381-ADEA 107.44 CAD
CAAA14381-AD CAAA14381-AE CAAA14381-AP
Selenium, plasma standard solution, Specpure®, Se 10,000 µg/ml, Thermo Scientific
Standards Cationic Standards Selenium Standards

Molecular Formula: Se in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134090

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR