Potassium, plasma standard solution, Specpure®, K 10,000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

14379-AD 14379-AE 14379-AP
CAAA14379-ADEA 127.66 CAD
CAAA14379-AD CAAA14379-AE CAAA14379-AP
Potassium, plasma standard solution, Specpure®, K 10,000 µg/ml, Thermo Scientific
Standards Cationic Standards Potassium Standards

Molecular Formula: KNO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00133776

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR