Hafnium, plasma standard solution, Specpure®, Hf 10,000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14374-AD
14374-AE
CAAA14374-ADEA
161.16
CAD
CAAA14374-AD
CAAA14374-AE
Hafnium, plasma standard solution, Specpure®, Hf 10,000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Hafnium Standards
Molecular Formula: HfCl2O in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011032
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...