Boron, Plasma Standard Solution, Specpure®, B 10,000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
14370-AD
14370-AE
14370-AP
CAAA14370-ADEA
116.29
CAD
CAAA14370-AD
CAAA14370-AE
CAAA14370-AP
Boron, Plasma Standard Solution, Specpure®, B 10,000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Boron Standards
Molecular Formula: H3BO3 in 1% NH4OH
Physical Form: Liquid
MDL No.: MFCD00134034
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...