Bis(triphenylphosphine)nickel(II) chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dichlorobis(triphenylphosphine)nickel(II), Bis(triphenylphosphine)nickel(II)chloride

13930-09 13930-18
CAAA13930-09EA 85.16 CAD
CAAA13930-09 CAAA13930-18
Bis(triphenylphosphine)nickel(II) chloride 98%
Bis(triphenylphosphine)nickel (II) chloride

Catalyst for cross-coupling of Grignard reagents, Hydrosilylations and hydrogenation, polymerizatron


Purity: 98% CAS No.: 14264-16-5 Molecular Formula: NiCl2[P(C6H5)3]2 Formula Weight: 654.18 Physical Form: Crystalline powder Sensitivity: Moisture Sensitive Appearance: Grey to green to black Odor: Odorless Melting Point: ca 225° dec. MDL No.: MFCD00009592


Catalyst for cross-coupling of Grignard reagents, Hydrosilylations and hydrogenation, polymerizatron

Formula: C₃₆H₃₀Cl₂NiP₂
MW: 656.19 g/mol
Melting Pt: ∼ 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00009592
CAS Number: 14264-16-5
EINECS: 238-154-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR