Gold, plasma standard solution, Specpure®, Au 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13881-AD
13881-AE
13881-AP
CAAA13881-ADEA
158
CAD
CAAA13881-AD
CAAA13881-AE
CAAA13881-AP
Gold, plasma standard solution, Specpure®, Au 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Gold Standards
Molecular Formula: Au in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00003436
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...