Erbium, plasma standard solution, Specpure®, Er 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13877-AD
13877-AE
CAAA13877-ADEA
84.85
CAD
CAAA13877-AD
CAAA13877-AE
CAAA13877-AP
Erbium, plasma standard solution, Specpure®, Er 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Erbium Standards
Molecular Formula: Er2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010987
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...