Zirconium, Plasma Standard Solution, Specpure®, Zr 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13875-AD
13875-AE
13875-AP
CAAA13875-ADEA
84.85
CAD
CAAA13875-AD
CAAA13875-AE
CAAA13875-AP
Zirconium, Plasma Standard Solution, Specpure®, Zr 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Zirconium Standards
Molecular Formula: ZrCl2O in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011303
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...