Strontium, Plasma Standard Solution, Specpure®, Sr 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13874-AD
13874-AE
13874-AP
CAAA13874-ADEA
84.85
CAD
CAAA13874-AD
CAAA13874-AE
CAAA13874-AP
Strontium, Plasma Standard Solution, Specpure®, Sr 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Strontium Standards
Molecular Formula: Sr(NO3)2 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134060
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...