Rubidium, Plasma Standard Solution, Specpure®, Rb 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13872-AD
13872-AE
CAAA13872-ADEA
80.11
CAD
CAAA13872-AD
CAAA13872-AE
CAAA13872-AP
Rubidium, Plasma Standard Solution, Specpure®, Rb 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Rubidium Standards
Molecular Formula: Rb2CO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134055
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...