Osmium, Plasma Standard Solution, Specpure®, Os 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
13871-AE 13871-AP 13871-AD 13871-AE 13871-AD 13871-AP
CAAA13871-APEA 927.46 CAD
CAAA13871-AP CAAA13871-AD CAAA13871-AE AA13871-AP AA13871-AD AA13871-AE
Osmium, Plasma Standard Solution, Specpure®, Os 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Osmium Standards
Molecular Formula: (NH4)2OsCl6 in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011147
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR