Lanthanum, Plasma Standard Solution, Specpure®, La 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13870-AD
13870-AE
13870-AP
CAAA13870-ADEA
81.69
CAD
CAAA13870-AD
CAAA13870-AE
CAAA13870-AP
Lanthanum, Plasma Standard Solution, Specpure®, La 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Lanthanum Standards
Molecular Formula: La2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011066
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...