Gallium, plasma standard solution, Specpure®, Ga 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13869-AD
13869-AE
13869-AP
CAAA13869-ADEA
80.26
CAD
CAAA13869-AD
CAAA13869-AE
CAAA13869-AP
Gallium, plasma standard solution, Specpure®, Ga 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Gallium Standards
Molecular Formula: Ga in 5% HNO3/tr. HCl
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134045
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...