Copper, plasma standard solution, Specpure®, Cu 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

13867-AD 13867-AE 13867-AP
CAAA13867-ADEA 85.79 CAD
CAAA13867-AD CAAA13867-AE CAAA13867-AP
Copper, plasma standard solution, Specpure®, Cu 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Copper Standards

Molecular Formula: Cu in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010965

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR