Potassium, plasma standard solution, Specpure®, K 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13866-AD
13866-AE
13866-AP
CAAA13866-ADEA
83.58
CAD
CAAA13866-AD
CAAA13866-AE
CAAA13866-AP
Potassium, plasma standard solution, Specpure®, K 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Potassium Standards
Molecular Formula: KNO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00133776
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...