Tin, Plasma Standard Solution, Specpure®, Sn 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13863-AD
13863-AE
13863-AP
CAAA13863-ADEA
91.48
CAD
CAAA13863-AD
CAAA13863-AE
CAAA13863-AP
Tin, Plasma Standard Solution, Specpure®, Sn 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Tin Standards
Molecular Formula: Sn in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00133862
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...