Magnesium, Plasma Standard Solution, Specpure®, Mg 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13861-AD
13861-AE
13861-AP
CAAA13861-ADEA
83.58
CAD
CAAA13861-AD
CAAA13861-AE
CAAA13861-AP
Magnesium, Plasma Standard Solution, Specpure®, Mg 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Magnesium Standards
Molecular Formula: Mg in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085308
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...