Boron, Plasma Standard Solution, Specpure®, B 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13859-AD
13859-AE
13859-AP
CAAA13859-ADEA
77.89
CAD
CAAA13859-AD
CAAA13859-AE
CAAA13859-AP
Boron, Plasma Standard Solution, Specpure®, B 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Boron Standards
Molecular Formula: H3BO3 in 1% NH4OH
Physical Form: Liquid
MDL No.: MFCD00134034
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...