Yttrium, Plasma Standard Solution, Specpure®, Y 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13855-AD
13855-AE
13855-AP
CAAA13855-ADEA
87.22
CAD
CAAA13855-AD
CAAA13855-AE
CAAA13855-AP
Yttrium, Plasma Standard Solution, Specpure®, Y 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Yttrium Standards
Molecular Formula: Y2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011468
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...