Calcium, Plasma Standard Solution, Specpure®, Ca 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13852-AD
13852-AE
13852-AP
CAAA13852-ADEA
87.69
CAD
CAAA13852-AD
CAAA13852-AE
CAAA13852-AP
Calcium, Plasma Standard Solution, Specpure®, Ca 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Calcium Standards
Molecular Formula: CaCO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085314
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...