Thallium, Plasma Standard Solution, Specpure®, Tl 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13851-AD
13851-AE
13851-AP
CAAA13851-ADEA
81.05
CAD
CAAA13851-AD
CAAA13851-AE
CAAA13851-AP
Thallium, Plasma Standard Solution, Specpure®, Tl 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Thallium Standards
Molecular Formula: Tl in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134063
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...